missing translation for 'onlineSavingsMsg'
Learn More
3-Phenylphenol, 90%, Thermo Scientific Chemicals Product Code.: 10647474

3-Phenylphenol, 90%, Thermo Scientific Chemicals

Product Code. 10647474
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10647474

Brand: Thermo Scientific Acros 417660050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-51-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002294
InChI Key UBXYXCRCOKCZIT-UHFFFAOYSA-N
Synonym 3-hydroxybiphenyl, 1,1'-biphenyl-3-ol, biphenyl-3-ol, m-phenylphenol, m-hydroxydiphenyl, 3-biphenylol, m-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, unii-zu11x47h4oShow MoreShow Less
PubChem CID 11381
ChEBI CHEBI:34338
IUPAC Name 3-phenylphenol
SMILES OC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 72.0°C to 80.0°C
Color Beige-Yellow to White
Boiling Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: organic solvents: ethanol,diethyl ether,chlorof,benzene,pyridine
Formula Weight 170.21
Percent Purity 90%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Phenylphenol, 90%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458257 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number417660050,417660250
Lot NumberA0454695 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number417660050,417660250
Lot NumberA0449878 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog Number417660050,417660250
Lot NumberA0408053 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number417660050,417660250
Lot NumberA0414917 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number417660050,417660250
compliance-icons
Product Identifier
  • 3-Phenylphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : [1,1'-Biphenyl]-3-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Phenylphenol, 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.