Learn More
Fisherbrand™ 96-Well Semi-Skirted PCR Plates
Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters
Brand:  Fisherbrand™ 14230246
114.00 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 11889261
  • 120.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Directly compatible with all major thermal cyclers, including ABI instruments—can be used in ABI 96-well sequencers without adapters
  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Raised-rim design
  • Sealable with adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced
Specifications
Specifications
Standard
Blue
Semi
300 μL
PCR, RT-PCR applications
Clear
8 x 12
PCR Microplate
Polypropylene
96
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ 96-Well Semi-Skirted PCR Plates > Blue