missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SDS Solution, 10%
GREENER_CHOICE

Filtered and ready to use

Brand:  Invitrogen™ 15553027

 View more versions of this product

Product Code. 11538896

  • 167.00 € / 400mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

UltraPure 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations.

  • Used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions
  • Sustainable packaging

DNA & RNA Purification & Analysis, DNA Extraction, Gene Expression Analysis & Genotyping, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Liquid
UltraPure™
SDS
SDS Solution
Nucleic Acid Gel Electrophoresis, Blotting, DNA Extraction
4 x 100 mL
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number445842-000 Certificate TypeCertificate of Analysis Date17/04/2025 Catalog Number
Lot Number440661-000 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number440661-000 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number
Lot Number2451914 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15553027
Lot Number2451910 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15553027
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ UltraPure™ SDS Solution, 10% > Quantity: 4 x 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.