missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene Measuring Scoop

Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.

Brand:  Azlon SWL030

 View more versions of this product

Product Code. 12521516

  • 51.15 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.

  • Made from food safe polypropylene
  • Top is flat for levelling off powders
  • Base is flat enabling scoops to stand horizontally
  • The capacity is clearly molded on each size
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Measuring Scoop
Handle
259 mm
Polypropylene
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ Polypropylene Measuring Scoop > Capacity: 250mL; Length: 259mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.