Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Brand: BRAND™ 701346
221.69 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
Description
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.

Specifications
Standard | |
Autoclavable | |
Polypropylene | |
96 | |
Flexible mat covers or adhesive sealing films to reduce sample evaporation and contamination | |
U-bottom |
8 x 12 | |
Deep-well Plate | |
0.5 mL | |
Round Bottom (U-well) | |
48 | |
48 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.