Learn More
BTX™ Tweezertrode™ Electrode Kit
Tweezertrode™ Electrode Kit features reusable, tweezer-style, in vivo electrodes suitable for drug or gene delivery in animals.
Brand: BTX™ 450166
Description
- Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel or platinum at the tip
- Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
- Can be easily cleaned
- Kits are supplied with BTX524 coaxial cable for connection to electroporation generators
Specifications
In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery. | |
Stainless Steel | |
Electrode Kit | |
Stainless Tweezertrode Kit, 10 mm | |
10 mm | |
Cable | |
0.47 in. | |
12 mm | |
1 Ea. | |
<0.04 in. to >0.79 in. | |
11.5 cm | |
Banana Cables |
ECM 830, Gemini X2, ECM 2001+ Legacy ECM 2001 Generator | |
BTX Electroporation | |
1000 V | |
0.39 in. | |
Circular | |
0.39 in. | |
10 mm | |
10 μsec. to 99 msec. in PBS | |
20 to 80% | |
<1 mm to >2 cm | |
Non-sterile | |
Square Wave |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Intended for Research use only. Not for use in diagnostic or clinical procedures.