Learn More
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit
Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.
Brand: Welch Ilmvac™ 402008
This item is not returnable.
View return policy
Description
- Available for a range of diaphragm vacuum pump models
- Provided individually
Specifications
Pump Models MP / MPC 101 Z, 201 E | |
Diaphragm, Valves And O-Rings. | |
1 Pack | |
Service Kit |
Product Suggestions
Customers who viewed this item also viewed.
Fisherbrand Vacuum pumps
File size: 5.6 MB
Product Content Correction
Product Title
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit > For Use With: Pump Models MP / MPC 101 Z, 201 E
Spot an opportunity for improvement?Share a Content Correction