missing translation for 'onlineSavingsMsg'
Learn More

Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196111000

20.43 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.



 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10153830

  • 21.51 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Manganese standard solution
7439-96-5, 7697-37-2, 7732-18-5
100 mL
1.02
MFCD00011111
997 to 1003 μg/mL of Mn (2+) in 2% HNO3 (20°C)
PWHULOQIROXLJO-UHFFFAOYSA-N
101.0°C
manganese
54.94
CHEBI:35154
AAS
1 mg/ml Mn in 2% HNO3
Plastic bottle
1.0200g/mL
Mn
Liquid
Colorless
-3.0°C
[Mn]
Soluble in water
23930
54.94
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468650 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number196115000,196111000
Lot NumberA0460083 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number196115000,196111000
Lot NumberA0451516 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number196115000,196111000
Lot NumberA0415809 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number196115000,196111000
Lot NumberA0374262 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number196115000,196111000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals > 100mL; Plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only