Learn More
Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips
Eliminate cell fragmentation caused by mechanical shearing with these wide orifice pipette tips, ideal for pipetting macromolecules.
35.51 € - 504.60 €
Product Code | Brand | Volume (Metric) | Sterility | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Format | Price | Quantity & Availability | |||||
11408499
![]() |
Thermo Scientific™
9405020 |
250 μL | Non-sterile | Bagged |
73.90 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
10053750
![]() |
Thermo Scientific™
9405050 |
1000 μL | Non-sterile | Bagged |
35.51 €
Pack of 400 |
In Stock
Log in to see stock. ![]() |
|||||
10329370
|
Thermo Scientific™
9405060 |
1000 μL | Non-sterile | Bulk | N/A | ||||||
10073560
![]() |
Thermo Scientific™
9405163 |
1000 μL | Sterile | Rack |
192.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||||
11428499
![]() |
Thermo Scientific™
9405120 |
250 μL | Non-sterile | Rack |
122.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||||
10349320
![]() |
Thermo Scientific™
9405123 |
250 μL | Sterile | Rack |
155.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||||
Description
Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips, ideal for pipetting macromolecules.
Key Features:
- Smooth, uniform inner surface of the tips prevents liquid retention
- Available as Sterile or Non-sterile option
- Packaging options: racked or bulk
Proven quality
- Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Tip Range
- Wide orifice tips available for volumes 250 and 1000 μL allowing better liquid handling for viscous liquids and sensitive cell samples
- Inside diameter of orifice is 1.1mm/1.8mm
Immediate traceability
- Each tip rack has ink-jetted lot and volume information
Note:
Availability may vary by country.
Specifications
Non-filtered | |
FinnTip Wide Orifice |