Learn More
Fisherbrand™ Cell Lifters
Disposable lifters quickly remove cell layers
Brand:  Fisherbrand™ 08100240
Product Code. 11577692
  • 387.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Disposable lifters quickly remove cell layers
The Fisherbrand Cell Lifter is used with dishes and cell culture clusters for quick removal of cell layers. The one-piece design and chiseled blade provide improved control while minimizing damage during the cell removal process.
  • Sterilized and individually wrapped in peel-open packaging
  • Polyethylene copolymer blade with angled and chiseled edge
  • Maintains even contact with growth area in culture dishes
Specifications
Specifications
Polyethylene
Removal of cell layers
Single-use
7 in.
17.78 cm
Cell Lifter
Sterile
Dishes and cell culture clusters
0.7 in.
2 cm
Angled and Chiseled
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Cell Lifters > Cell lifter