Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Brand:  SCAT™ 107048
 View more versions of this product
Product Code. 15491793
Description
Description
SCAT fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Specifications
Fitting
1.6 mm
Fitting with Integrated Ferrule (1.6 mm OD)
PFA
Colorless
Solvent Handling
Capillary (1.6 mm OD)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCAT™ Fittings for HPLC Systems > Fittings; Color: Colorless; For Use With: Capillary (1.6 mm OD); Includes: Fitting with Integrated Ferrule (1.6 mm OD); Type: Fitting