Learn More
Gibco™ Trypsin (2.5%), no phenol red

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas
Brand: Gibco™ 15090046
25.69 EUR valid until 2025-12-31
Use promo code "24335" to get your promotional price.
Description
Due to its digestive strength, trypsin solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation; concentration required for dissociation varies with cell type and experimental requirements.
- Solutions tested for pH, osmolality, sterility, and performance
- Prior to manufacturing, the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
- Documented Traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency
Sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Compliance
Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications
No EDTA | |
Mammalian Cells | |
10 X | |
Cell Culture Dissociation Reagent | |
No Phenol Red | |
7.1 to 8 | |
100 mL | |
Animal Origin |
Storage conditions: -5°C to -20°C Shelf life: 24 months from date of manufacture |
|
Animal Origin | |
Liquid | |
Sterile-filtered | |
270 - 320 mOsm/kg | |
Gibco™ | |
Wet Ice | |
In vitro Bioassay |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- TRYPSIN, 2.5%, 10X
Signal Word
- Danger||Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trypsin
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2428460 | Certificate of Analysis | 21/02/2025 | 150901000,150900250,10406800,15090046,150905000 |
2428462 | Certificate of Analysis | 21/02/2025 | 150901000,150900250,10406800,15090046,150905000 |
2365735 | Certificate of Analysis | 21/02/2025 | 150901000,150900250,10406800,15090046,150905000 |
2569831 | Certificate of Analysis | 21/02/2025 | 150901000,150900250,10406800,15090046,150905000 |
2677672 | Certificate of Analysis | 21/02/2025 | 150901000,150900250,10406800,15090046,150905000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.