Learn More
Gilson™ GmCLab Fixed-Speed Mini-Centrifuge
Gilson™ GmCLab Fixed-Speed Mini-Centrifuge provides quick separation of particles from supernatant in solid-liquid mixture, or spins down liquids to bottom of microtube.
Brand: Gilson™ F110733
Description
- Compact design: Easy integration on busy workstations with its small footprint
- Robustness: Robust lid hinge for long lasting spin operation
- Ease of use: Quick spin operation by closing and opening the lid
- Easy access to tubes with fully opening lid
- No tools required: Pinch-type connector to quickly exchange rotors
- Emergency brake stops the centrifuge immediately if any tube opens accidently and splashes liquid inside
- Maintenance free: No budget needed for servicing
- Lid, body, rotors and bottom of module are chemically compatible with most solvents, acidic and alkaline buffers
Specifications
8 x 1.5/2 mL with appropriate rotors | |
No | |
No |
Product Suggestions
Customers who viewed this item also viewed