Learn More
Unity Lab Services™ Incubator Requalification Service
Requalification Services for Incubator (documentation included)
Brand:  Unity Lab Services™ RQINC
Product Code. 30178680
  • 1532.00 € / 1 unit
Explore more special offers
Description
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Incubator
Requalification Service
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Unity Lab Services™ Incubator Requalification Service > Requalification Service