Learn More
LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
For water quality testing
Brand: LaMotte™ 99532-34
Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Water quality testing | |
50 Strips/Vial | |
Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
Chlorine dioxide | |
0 to 500 ppm | |
Single Factor Test Strip |