missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar with Salt

Isolate and presumptively identify coliforms and intestinal pathogens in water, dairy and biological samples with this differential, ready-to-use medium.

Brand:  Thermo Scientific™ PO0149A

Product Code. 12982578

  • 16.35 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Isolate and presumptively identify coliforms and intestinal pathogens in water, dairy and biological samples with Thermo Scientific™ Oxoid™ MacConkey Agar with Salt. Owing to its ability to support the growth of pathogenic Gram-positive cocci as well as Enterobacteriaceae, the medium is particularly recommended for the cultivation pathogens from clinical specimens. MacConkey Agar with Salt also provides a number of diagnostic differentiators such as bile tolerance, colony morphology and chromogenesis.

Use MacConkey Agar with Salt as a differential medium for the isolation of coliforms and intestinal pathogens in water, dairy and biological samples.

  • Cultivate pathogens from clinical specimens and dairy products.
  • Conduct water testing for coliforms and pathogens.
  • Use in parallel with other selective indicator media such as Desoxycholate Citrate Agar, Bismuth Sulphite Agar, Brilliant Green Agar, and a non-selective medium such as Blood Agar.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dark Red
MacConkey Agar with Salt
United Kingdom
19.5g±1.0.g
10/Pk.
2°C to 10°C
90mm Monoplate
Agar
10 x 90mm plates, film wrapped
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4500334 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberPO0149A
Lot Number4499852 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPO0149A
Lot Number4497262 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberPO0149A
Lot Number4495602 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberPO0149A
Lot Number4492343 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPO0149A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MacConkey Agar with Salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.