Learn More
Applied Biosystems™ MicroAmp™ 3 x 32-Well Retainers
Designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System
Brand: Applied Biosystems™ 4481669
31.07 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- The Applied Biosystems MicroAmp 3 x 32-Well Retainer is specifically designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System.
- The retainer provides support and allows pressure to be distributed evenly over tubes and strips to ensure a proper seal and prevent the tubes and strips from being crushed or deformed.
- Its use is recommended at all times when working with MicroAmp reaction tubes and tube strips.
- Simply place the retainer on the sample block, then load the tubes or strips.
Specifications
Certified DNA/RNase Free | |
MicroAmp | |
Retainer | |
PCR | |
1 retainer |
Store at room temperature. |
|
Black | |
ProFlex 3 x 32-Well PCR System | |
32-Well Plate | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.