Learn More
MP Biomedicals™ Yeast, Torula, Powder
Source: Candida utilis
Brand: MP Biomedicals™ 0290308525
Description
Applications:
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers. It is also used in pet foods, in beekeeping, and as an attractant in traps designed to control olive flies

Specifications
Animal Diet | |
Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies | |
15°C to 30°C |
Powder | |
11.33 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.