Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
GREENER_CHOICE
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
Brand:  Mettler Toledo™ 30111138
121.79 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15322275
  • 132.00 € / Each
Explore more special offers
Description
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
  • NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
  • Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
  • A test certificate and detailed label with a temperature table comes with every box
  • All SDS and labels contain information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIST/DIN Buffer pH 9.180 sachets 30x20mL
pH measurement, electrode calibration
Sachet
30 x 20 mL
pH Electrode
Yes
9.180 (at 25°C)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets > NIST/DIN Buffer pH 9.180 sachets 30x20mL