Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.
Brand: Thermo Scientific™ B2
Description
Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.
The EasyCast systems feature leakproof casting that allows a gel to be cast and run in the same chamber with no tape or additional parts required. The EasyCast B2's longer gel length and the 2 comb slots provide the flexibility of running 8 to 48 samples on 1 gel. The optional multi load tray can increase the maximum number of samples to 108 in an 8 or 12 multichannel pipette format.
- 2 comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
Buffer chamber, SuperSafe™ lid with attached power supply leads, EasyCast gasketed U.V. Transmissible (UVT) gel tray, 2 combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick

Specifications
CE marked | |
Owl | |
EasyCast | |
800 mL | |
9.63 in. | |
14 cm | |
18 cm | |
3.75 in. | |
150 V |
Mini | |
Mini Gel Electrophoresis System | |
8 to 48 | |
9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm) | |
24.5 cm | |
7.06 in. | |
12 cm | |
9.5 cm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.