missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock

HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.

Brand:  Fisherbrand™ 20 36 2402

79.82 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 12941031

  • 111.28 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Lock
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
20 mL
Luer Lock
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock > Volume: 20mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.