Learn More
Fisherbrand™ Aluminum Weighing Dishes
Aluminum dishes for weighing and dispensing
Brand:  Fisherbrand™ 08732101
 View more versions of this product
Product Code. 10297593
  • 33.38 € / Pack of 144
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.
With Fluted sides
  • Also can serve as evaporating dishes or dust covers Lightweight, yet sturdy Convenient finger-grip handle can be etched prior to weighing for identification
With Smooth sides
  • Lightweight dishes with flanged rims
  • Slightly sloping sides facilitate stacking
  • Especially useful for materials that cannot easily be cleaned from dish surface
Specifications
Specifications
Flat
Aluminum
Weighing Dish
17 mm
62 mm
42 mL
Fluted Sides
For weighing and dispensing
51 mm
51 mm (Inner Base), 62 mm (Inner Top)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Aluminum Weighing Dishes > With Fluted Sides; Capacity: 1.375 fl. oz.