missing translation for 'onlineSavingsMsg'
Learn More

Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle

Features a robust pail handle for easy transportation and a wide aperture for easy insertion of larger items. Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.

Brand:  Daniels Healthcare DD476YL

Product Code. 12953495

  • 28.95 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications

Specifications

9.27 L
Lid
242 mm
Autoclavable
For easy transportation
BS7320:1990, UN3291, AFNOR, NFX 30-500
Yellow
280 mm
275 mm
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle > Dimensions: 280L x 250mmH; Capacity: 11.5L; Color: yellow

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.