Learn More
SPDP, MedChemExpress
MedChemExpress SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
Brand:  Medchem Express HY-100216
Encompass_Preferred
 View more versions of this product
Product Code. 16471516
  • 1344.60 € / 500mg
Explore more special offers
Description
Description
SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
Specifications
Specifications
SPDP
White
C12H12N2O4S2
SPDP Crosslinker
O=C(ON1C(CCC1=O)=O)CCSSC2=NC=CC=C2
312.36
Research
68181-17-9
Research
500 mg
DMSO : 155 mg/mL (496.22 mM; Need ultrasonic)
312.36
97.0%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SPDP, MedChemExpress