missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mini-Magnifier

Enlarges objects for easier viewing. Fisherbrand™ Mini-Magnifier swivels for comfortable observation.

Brand:  Fisherbrand™ HS15092D

Product Code. 12816355

  • 112.00 € / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • The light box is ideal for quick screening of 96 well plates, Petri dishes, electrophoresis gels, slides and transparencies.
  • Requires 2 x AA batteries (included).
  • Swiveling, molded lens provides 1.75X magnification.
Specifications

Specifications

Red Frame
8.1 in.
Frame and Magnifier
16.1 cm
1 oz.
5.3 in.
1.75x
Mini-Light Box
20.7 cm
6.3 in.
1 Ea.
0.45 kg
13.5 cm
Magnifier
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Mini-Magnifier > Magnification: 1.75X

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.