Learn More
Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
Brand:  Fisherbrand™ FB71281
 View more versions of this product
Product Code. 10637584
  • 76.80 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Provide extraordinary flexibility
  • Can be combined with a wide variety of different grid inserts
Specifications
Specifications
White
Cryo Box
6 mm to 28.5 mm Diameter Tubes
133 x 133 x 100 mm
Cardboard
Tubes from dia. 6mm to dia. 28.5mm
10 Pack
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating > Height: 100mm; Color: White; Format: 133 x 133mm