Learn More
Thermo Scientific™ RNA Gel Loading Dye (2X)
Prepare RiboRuler RNA ladders and RNA samples for electrophoresis on agarose or polyacrylamide gels using Thermo Scientific™ 2X RNA Loading Dye.
Brand: Thermo Scientific™ R0641
29.80 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
2X RNA Loading Dye is recommended for preparation of RiboRuler RNA ladders and RNA samples for electrophoresis on agarose or polyacrylamide gels. It contains the tracking dyes bromophenol blue and xylene cyanol FF as well as the intercalating dye ethidium bromide. In most denaturing agarose gel systems, bromophenol blue migrates slightly faster than human 5S rRNA, whereas xylene cyanol FF migrates slightly slower than 18S rRNA.
2X RNA Loading Dye contains the denaturing agent formamide, which allows RNA fragments to separate according to size even during non-denaturing electrophoresis. Formamide also stabilizes RNA.
Highlights
- Two-color tracking of DNA, RNA migration during electrophoresis
- No DNA, RNA masking during gel exposure to UV light
- Denaturation of DNA, RNA due to the presence of formamide

Specifications
2X | |
Acrylamide Gels, Agarose Gels | |
RNA Gel Loading Dye |
RNA | |
Bromophenol Blue, Xylene Cyanol FF (XCFF) | |
1 mL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.