missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Cutlery
GREENER_CHOICE

Packaged individually in tamper evident zip bags. Gosselin™ Sterile Cutlery will help handle primary samples with limited cross contamination risks.

Brand:  Corning™ CUIP-01

 View more versions of this product

Product Code. 11847363

  • 528.37 € / Pack of 1000
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Resistant Polystyrene (PS) cutlery
  • Complete range
  • Sterile sample handling, single bag
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spoon
White
125 mm
Large
0.067 oz.
Polystyrene
4.9 in.
1000/Cs.
Sterile
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Sterile Cutlery > 1/Bag

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.