Learn More
BRAND™ Multi-Channel Micropipette Transferpette™ S -8, adjustable, DE-M

Efficient: one-hand operation. Flexible: ergonomic design for all hand sizes. Reliable: volume-change protection prevents accidental adjustments.
Brand: BRAND™ 705910
696.35 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
The Transferpette™ S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette™ S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.
- The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment.
- The Transferpette™ S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools.
- A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
- Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.

Specifications
DIN EN 285 | |
Autoclavable | |
Transferpette S | |
Pipette | |
15 to 40°C | |
23.7 cm | |
12.3 cm |
Transferpette™ S-8, adjustable volume, DE-M, M8-200, 20-200 μL | |
Polypropylene | |
Non-sterile | |
20 to 200 ÎĽL | |
36 months | |
14 cm |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Brand Transferpette S (EN)
Your input is important to us. Please complete this form to provide feedback related to the content on this product.