Learn More
Fisher Chemical 25KG SODIUM HYDROXIDE
Brand: Fisher Chemical S/4800/70
100.67 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
25KG SODIUM HYDROXIDE
Description

Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2530961 | Certificate of Analysis | 21/03/2025 | S/4800/70 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.