Learn More
Tuttnauer 65L Benchtop Autoclave
The 65L Benchtop Autoclave has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Brand: Tuttnauer 3850EAP-D
Description
EL - D Line models:
Tuttnauer benchtop laboratory autoclaves are front loading autoclaves available in chamber sizes from 28 to 160 liters. EL models have an advanced multi-color control panel and a chamber made of 316L stainless steel. Tuttnauer laboratory autoclaves have been designed to provide high quality repeatable performance and accountability for a wide range of applications used in modern laboratories.
- Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
- Benchtop lab autoclaves are designed to save space on your laboratory workbench.
Specifications
14.3 gal. | |
50 cm | |
20 in. | |
30.6 in. | |
3850EAP-D | |
221°F to 279°F | |
Drying (Not Vacuum) | |
Non-sterile |
65 L | |
15.2 in. | |
38 cm | |
765 mm | |
Benchtop Autoclave | |
105°C to 137°C | |
316L Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Accessories and Services
-
Tuttnauer Isothermal/Disinfection (for Agar Preparation) Disenfection (agar prep) Product Code: 17809538
769.00 € / Each -
Tuttnauer Additional Year Warranty Additional one-year warranty Product Code: 17839548
1745.00 € / Each -
Tuttnauer IQ/OQ/PQ Documentation IQ/OQ/PQ Documentation Product Code: 17839568
2242.50 € / Each -
Tuttnauer Wheels Swivel 4 Wheels (2 Brakes) Wheels Swivel 4 Wheels (2 with bra Product Code: 17869548
484.00 € / Each -
Tuttnauer Printer for All Models (External) Printer for all models Product Code: 17879538
959.00 € / Each -
Tuttnauer Internal Printer for D Line Autoclaves Internal printer for 38XX/50XX Product Code: 17889538
1145.00 € / Each
Your input is important to us. Please complete this form to provide feedback related to the content on this product.