Learn More
IKA™ C1 Calorimeter Package with C1.10 Combustion Chamber
The C1 Calorimeter Package with C1.10 Combustion Chamber combines a compact design with a high degree of automation, featuring a light and easily retractable combustion chamber.
Brand:  IKA™ 0010005635
 View more versions of this product
Product Code. 17257479
  • 22935.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a high degree of automation
  • Compact design offers a small footprint
  • Operates in accordance with DIN 51900 and ISO 1928
  • PCI / PCS measurement analysis follows classic isoperibolic methodology
  • Light and easily retractable combustion chamber
  • Compatible with IKA™ C6040 Calwin calorimeter software (sold separately), which allows additional extensions and adaptations, such as the management of measurement data and LIMS
  • Includes wear parts and consumables for the first 500 tests including 25 calibrations
  • C1.10 combustion chamber is fitted with a large C5010.5 crucible support and a large C6 quartz crucible
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calorimeter
50/60 Hz
Up to 40,000 J
100/240 V
DIN 51900, ISO 1928
C1 Calorimeter, C1.10 Combustion Chamber, C1.20 Heater
20°C to 30°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ C1 Calorimeter Package with C1.10 Combustion Chamber > C1/C1.10 Calorimeter Package; Includes: C1 Calorimeter, C1.10 Combustion Chamber, C1.20 Heater