Learn More
HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™
Brand: Honeywell Fluka 34801-6X1L
480.58 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
View more versions of this product
Description

Specifications
HYDRANAL™-Titrant 5 | |
11°C (51.8°F) | |
UN1230 |
0.85g/mL | |
6 x 1 L |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- HYDRANAL Titrant 5
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, iodine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.