Learn More
Kimberly-Clark Professional™ Kleenex™ Facial Tissues, 2 Ply Boxed Tissues

Soft and luxurious facial tissues provide elevated care. Packed in a tissue box for a stylish, professional image. Each box contains high quality, soft, strong and absorbent 2 ply facial tissues that are produced from FSC certified fibres.
Brand: Kimberly-Clark Professional™ 8834
46.82 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Each individual box of tissues contains 2 ply tissues
- Tissues are dispensed from a stylish tissue box for a high-quality image
- Soft facial tissues ideal for use in homes, offices, and washrooms
- Produced from FSC (Forest Stewardship Council) certified wood
- Uncompromised care delivering added workplace hygiene from the trusted Kleenex™ brand

Specifications
Facial Tissue | |
30.8 cm | |
23.4 cm | |
FSC certified | |
20 x 20.6 cm | |
90 Sheets x 12 Dispenser Cubes |
12.2 in. | |
9.2 in. | |
Paper | |
White | |
Offices, Fitness Clubs, Hotels, Large Offices, Contract Cleaners, Washrooms | |
2-ply |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.