Learn More
Thermo Scientific™ Oxoid™ Yeast Extract Powder
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
Brand: Thermo Scientific™ LP0021B
Description
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
- Recommended for plate count of organisms in water
- Increases yield of organisms of many media or fermentation broths

Specifications
Yeast Extract Powder | |
Powder | |
Microbiology Media | |
500 g |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4489687 | Certificate of Analysis | 13/03/2025 | LP0021B |
4448128-02 | Certificate of Analysis | 16/04/2024 | LP0021B |
4439757-02 | Certificate of Analysis | 15/04/2024 | LP0021B |
4445142-02 | Certificate of Analysis | 25/03/2024 | LP0021B |
4443038-02 | Certificate of Analysis | 25/03/2024 | LP0021B |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.